Learn More
Fisherbrand™ Traceable™ Refrigerator/Freezer Plus Thermometer
Accurately monitors temperatures in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor
Brand: Fisherbrand™
Code : 50
Additional Details : Weight : 0.16740kg
Description
Fisher Scientific™ Traceable™ Refrigerator/Freezer Plus Thermometer accurately monitors temperatures in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor.
With 5mL round bottle. Vaccine bottle mimics vaccine temperature and fits in vaccine tray.- Probe is sealed in glycol solution
- Bottle insulates sensor from rapid temperature changes when refrigerator door is opened
- Monitors high/low readings overnight, on weekend or for any time period - a significant advantage over reading-only glass thermometers
- Temperature range is -58 to 158°F and -50 to +70°C with a resolution of 0.1 degree and an accuracy of +/- 0.5C
- Minimum/maximum memory
- High and low settable alarms
Spécification
ISO 17025, A2LA, NIST | |
3.25 x 2.5 x 0.6 in. (8.3 x 6.35 x 1.5 cm) | |
0.6 in. | |
Velcro™, magnetic strips, traceable certificate and AAA battery | |
11.2 ft. | |
3.4 m | |
ABS Plastic | |
4 oz. | |
2.5 in. | |
158°F | |
-58°F | |
-58°F to +158°F | |
Monitoring Temperature in Freezer | |
6.35 cm |
0.18 in. | |
Monitoring temperature in freezers, water baths, heating blocks, incubators and refrigerators | |
1.5 cm | |
120 in. | |
83 mm | |
3.17 cm | |
0.1°C | |
110 g | |
3.25 in. | |
70°C | |
-50°C | |
-50°C to +70°C | |
±0.5°C (±0.9°F) (between 0-50°C) |