Learn More
Fisherbrand™ Traceable™ Visual Alarm Timer
Ideal for repetitive tests
Brand:  Fisherbrand™ 93678-86
Product Code. 11719805
  • £22.05 / Each
Explore more special offers
Includes: AAA batteries (2), Integrated magnet, Stand, Clip
Description
Description
Ideal for repetitive tests. Fisher Scientific™ Traceable™ Visual Alarm Timer has a switch which allows user to select either an audible alarm or bright flashing LED visual alarm.
  • Counts down from 9999 minutes in one-minute increments
  • Countdown resolution is 1 second for times under 99 minutes
  • Automatic memory resets the display to the last programmed time
  • Thru-zero feature allows counting down, alarming, then counting up
  • Digits are 1cm (3/8 in.) high and readable from 3M (10 ft.)
Specifications
Specifications
Benchtop Timer
Audible/Visible
1 second to 1 minute
AAA
For repetative tests
Timing Capacity: 9999 min.
0.0001
LED
Two AAA batteries, integrated magnet, stand, clip
NIST