missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Made with precision machined aluminum rod for strength and durability
Has three knobs:
- Knob one: Raises and lowers stirrer
- Knob two: Locks stirrer on support rod
- Knob three: Controls swivel setting
Specifications
Stirring stands from 12mm to 19mm dia. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction