missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Vessel Holders for Heavy-duty Vortex Mixers

For use with heavy-duty vortex mixers.

Brand:  Fisherbrand™ 80863050

 View more versions of this product

Product Code. 15579024

  • £44.90 / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Rubber holder secures 125 and 250mL Erlenmeyer flasks, or 500 and 1000mL Erlenmeyer flasks
  • Requires Universal Holder

Specifications

Specifications

125 to 250 mL
Rubber
Grip mat
Universal holder
Small vessel holder
Heavy-duty vortex mixer
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers