Learn More
Fisherbrand™ Vortex Mixer Head Cover
For use with Fisherbrand vortex mixers.
Brand:  Fisherbrand™ 80863000
Product Code. 15766949
  • £45.14 / Each
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
  • Rubber head (76mm) 02-215-382 is used with 76mm head cover 02-215-383 for mixing beakers, flasks, and irregular shaped objects
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Head Cover
Vortex Mixer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Vortex Mixer Head Cover > Head Cover