missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Vortex Mixer Accessories Kits
For use with Fisherbrand vortex mixers.
£94.80 - £247.00
Product Code | Brand | Product Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Includes | Price | Quantity & Availability | ||||
15726949
|
Fisherbrand™
|
Foam Microtube and Microwell Plate Kit with Retainer | One microtube holder, one microwell plate holder, and one insert retainer |
£137.00
Each |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
15756939
|
Fisherbrand™
|
Foam Tube Holder Kit with Retainer | Holder for 9 to 13mm, 14 to 19mm and 20 to 25mm tubes, flat form insert, vessel harness, insert retainer |
£247.00
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
15766959
|
Fisherbrand™
|
Microtube Kit | Mixes (48) 0.25 to 2mL micro tubes; foam insert and insert retainer |
£94.80
Each |
Estimated Shipment: 09-10-2024
Log in to see stock. |
||||
Description
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
Foam Microtube and Microwell Plate Holder Kit with Retainer
- Includes one microtube holder, one microwell plate holder, and one insert retainer (Cat. Nos. 02-215-386, -387, -391)
Foam Tube Holder Kit with Retainer
- Iincludes 3 tube holders (for 9-13mm, 14-19mm, and 20-25mm tubes), one flat foam insert, two vessel harnesses and one insert retainer (Cat. Nos. 02-215-393, -394, -395, -397, -392, -391)
- Mixes (48) 0.25 to 2mL micro tubes; includes foam insert and insert retainer
- For mixing 96-well plates or deep well blocks; includes foam insert and insert retainer
Specifications
Vortex mixer |