missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Wall-Mount Clock, Calendar, and Thermometer

High-tech design looks great in any science classroom or office.

Brand:  Fisherbrand™ 90010-55

Product Code. 11795833

  • £206.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Two AAA batteries, wall mount

Description

Description

  • Read the time, date and temperature with one glance at the large 6.5 × 3.75 in. display
  • Time can be displayed in 12-hour AM/PM or 24-hour international time
  • Temperature is °C/°F selectable
  • Calendar displays month, date and day of the week

Certifications

Individually serial-numbered Traceable™ Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.

Specifications

Specifications

Calendar Thermometer Clock
1 sec.
AAA
-5°C to +50°C
9.25 x 10 in. (23.5 x 25.4 cm)
23.5 cm
25.4 cm
2.54 cm
0.53 kg
High tech design looks great in any science classroom or office
ISO 17025, A2LA, NIST
0.0001
LCD
23°F to 122°F
0.5°
9.25 in.
10 in.
1 in.
19 oz.
Three AAA batteries, wall mount
Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.