missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Wellwash™ and Wellwash™ Versa™ Microplate Strip Washer: Spare Bottles
Accessories for Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
£172.00 - £215.00
Specifications
Type | Storage Bottle |
---|---|
Includes | Solid cap |
For Use With (Equipment) | Wellwash Microplate Strip Washer, Wellwash Versa Microplate Strip Washer |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11717996
|
Thermo Scientific™
N10822 |
4 L |
£215.00
Each |
Estimated Shipment: 09-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11707996
|
Thermo Scientific™
N10821 |
2 L |
£172.00
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
For use with Fisher Scientific Wellwash and Wellwash Versa Microplate Strip Washer
Subsitute bottles of different sizes for added convenience
ELISA-based assays, cell washing
Specifications
Storage Bottle | |
Solid cap | |
Wellwash Microplate Strip Washer, Wellwash Versa Microplate Strip Washer |