missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Horizontal Large Gel Systems: Microplate Combs

For large gel systems

£49.50 - £54.55

Specifications

For Use With (Equipment) Owl A2 and A5 Large Gel Electrophoresis System
Type Micro Well Comb
Format Microplate
View More Specs

Products 4
Product Code Brand Teeth Count Thickness (Metric) Teeth Volume (Fill) Metric Price Quantity & Availability  
Product Code Brand Teeth Count Thickness (Metric) Teeth Volume (Fill) Metric Price Quantity & Availability  
11787125
View Documents
Fisherbrand™
A2-MTC
21 0.1 cm 35 μL
£50.70
Each
Estimated Shipment: 07-03-2025
to see stock.
Add to basket
 
12483657
View Documents
Fisherbrand™
A2-MT2C
42 0.1 cm 13 μL
£49.50
Each
Estimated Shipment: 07-03-2025
to see stock.
Add to basket
 
11955886
View Documents
Fisherbrand™
A2-MT2D
42 0.15 cm 20 μL
£54.55
Each
In Stock
to see stock.
Add to basket
 
11797125
View Documents
Fisherbrand™
A2-MTD
21 0.15 cm 50 μL
£49.50
Each
Estimated Shipment: 04-04-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

Owl A2 and A5 Large Gel Electrophoresis System
Micro Well Comb
Microplate
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.