missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Pouches for FB-GC10-1 Vertical Gel Caster

Product Code. p-143265
Click to view available options
Quantity:
100/Pk.
25/Pk.
Unit Size:
Pack of 100
Pack of 25
This item is not returnable. View return policy

Product Code. 11982975

Brand: Fisherbrand™ GP4100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Pouches for use with FB-GC10-1 Vertical Gel Caster.

  • Dimensions: 10L × 10cmW

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Owl JGC-4 Gel Casting System
Type Gel Pouch
Quantity 100/Pk.
Length (Metric) 10 cm
Width (Metric) 10 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.