missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cryogenic Dots, 0.5 in. Diameter
GREENER_CHOICE

Intended uses of cryostorage with liquid nitrogen, refrigeration and ultra-low temperature freezer applications.

Brand:  Fisherbrand™ 11010218

 View more versions of this product

Product Code. 17897201

  • £50.50 / 20 sheets
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The labels can be printed on and will not bleed. Perfect for distinguishing amongst various specimens.
Specifications

Specifications

Red
Non-sterile
0.5 in.
Dot label sheet
192
12.7 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers