missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 1536-Well Polystyrene Plates
Engineered for high throughput screening, allowing end users to screen four times as many samples in one plates
£289.93 - £314.18
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Array | 32 x 48 |
Product Type | 1536-Well Cell Culture Multiwell Plates |
Material | Polystyrene |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
13595440
|
Fisherbrand™
|
White |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13575440
|
Fisherbrand™
|
Clear |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
13585440
|
Fisherbrand™
|
Black |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Optimized for robotic instruments, plate readers and liquid handling systems
- Highly specified polystyrene
- Available in black, white and clear
- Low plate height optimized for stacking efficiency
- Working volume range: 1–10lμL/well
- Untreated
- Non-sterile
Order Info
Lids sold separately
Specifications
Standard | |
32 x 48 | |
Polystyrene | |
1 to 101 μL/well | |
Non-treated | |
For low volume assays such as colorimetric, enzymatic, cloning, replication and array, as well as storage |
Without Lid | |
1536-Well Cell Culture Multiwell Plates | |
1536 | |
Non-sterile | |
Flat Bottom (F-well) |