Learn More
Fisherbrand™ 3/8 in. Dot Labels
GREENER_CHOICE
Product Code. 17857221 Shop All Fisherbrand Products
Click to view available options
Color:
Blue
Brown
Gold
Green
Lavender
Orange
Pink
Purple
Red
Silver
Yellow
Unit Size:
Each
Product Code. 17857221
Only null left
Add to basket
Add to basket
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196°C (-311°F). Our cryo labels can be printed/written on and will not bleed, smudge or smear–perfect for identifying specimens after long periods of cryogenic storage.
  • Labels can be printed/written on and will not bleed, smudge or smear - perfect for identifying specimens after long periods of cryogenic storage
Specifications
Color Purple
Format Roll
Product Type Dot Label
Sterility Non-sterile
Quantity 1000/Roll
Diameter (English) 0.37 in.
Diameter (Metric) 9.55 mm