missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 384-Well Polystyrene Plates
Shop All Fisherbrand Products
Click to view available options
Color:
Black
Clear
White
Well Shape:
Rounded Square
Triangular, Flat Bottom (F-well)
Unit Size:
Pack of 100
Pack of 90
Description
Optimized for robotic instruments, plate readers and liquid handling systems.
- Highly specified polystyrene
- Ideal for low-volume assays such as colorimetric, enzymatic, cloning, replication, and array, as well as storage
- Flat well bottom surface provides greater area for binding
- Working volume: 10–100μL
- 384-well microplates conserve samples and reagents, providing cost savings and greater screening productivity
- Untreated
- Non-sterile
Order Info
Accommodate 12-565-511 lid (sold separately).
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 16 x 24 |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 10 to 100 μL |
| Sterility | Non-sterile |
| Surface Treatment | Non-treated |
| Well Shape | Rounded Square |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction