Learn More
Fisherbrand™ 384-Well Polystyrene Plates
Process four times as many samples as standard 96-well plates
Brand:  Fisherbrand™
333.56 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 7.39000kg
 View more versions of this product
Product Code. 13565440
  • £417.00 / Pack of 100
Estimated Shipment: 19-11-2024
to see stock.
Add to basket
Description
Description
Optimized for robotic instruments, plate readers and liquid handling systems.
  • Highly specified polystyrene
  • Ideal for low-volume assays such as colorimetric, enzymatic, cloning, replication, and array, as well as storage
  • Flat well bottom surface provides greater area for binding
  • Working volume: 10–100μL
  • 384-well microplates conserve samples and reagents, providing cost savings and greater screening productivity
  • Untreated
  • Non-sterile
Order Info
Accommodate 12-565-511 lid (sold separately).
Specifications
Specifications
Standard
16 x 24
384-Well Microplate
384
Non-sterile
Triangular, Flat Bottom (F-well)
180/Cs.
Without Lid
Clear
Polystyrene
10 to 100 μL
Non-treated
For low volume assays such as colorimetric, enzymatic, cloning, replication and array, as well as storage