Learn More
Fisherbrand™ 384-Well ShallowWell Polypropylene Microplates
Small volume applications
Brand:  Fisherbrand™
392.46 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Code : NEW
Additional Details : Weight : 3.80000kg
 View more versions of this product
Product Code. 13595450
  • £611.00 / Pack of 100
In Stock
to see stock.
Add to basket
Description
Description
Feature a shared-wall design which yields increased well volume
  • Automation compatible, ANSI standard
  • Low-binding and chemically resistant surface
  • Natural plates for storage or colorimetric assays
  • White plates for luminescent assays, including SPA
  • Black plates for fluorescent assays
  • Round bottom wells for optimal sample recovery
  • Rounded square wells minimize wicking (capillary action)
  • Working volume range: 2 to 35μL/well
Order Info
Lids not included
Specifications
Specifications
ANSI
16 x 24
Polypropylene
384
Round, Conical Bottom (V-well)
Standard
Black
120 μL
Non-sterile
Fluorescent assays