missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Easy-Squeeze Wash Bottles, 6/Cs.
GREENER_CHOICE

Product Code. 11705243
Click to view available options
Color:
Assorted
Blue
Green
Natural
Red
Yellow
For Use With (Application):
Acetone
Bleach
Cleaning assortment pack
Deionized Water
Distilled Water
Ethanol
Isopropanol
Methanol
Saline
Soap
Solvent assortment pack
Universal
Water
Wash bottles in this range that are prelabelled for intended usage with specific hazardous substances are also prelabelled with the appropriate GHS hazard warning symbols.
 
Wash bottles in this range that are prelabelled for intended usage with specific hazardous substances are also prelabelled with the appropriate GHS hazard warning symbols.
 

Includes

Come with colored polypropylene closures with PTFE membrane (except 02-897-2, which has an HDPE closure).

Easy to fill and ready to use

  • Wide mouth
  • Translucent and sputter-free
  • With easy-to-read color-coded silk-screened labels
  • Tips can be cut back to increase flow
  • When not in use, bottles relieve internal pressure without dripping

Label information includes:

  • Chemical name
  • National Fire Protection Association (NFPA) codes and symbols
  • Chemical Abstracts Service (CAS) number
  • Hazard codes
  • Laboratory protective equipment information
  • Target organ information
  • Recommended fill level
  • LDPE recycling code
  • Bottle volume capacity “For Dispensing Only, Not Recommended For Storage” statement

Not recommended for use above 60°C (140°F).

Order Info

Available in cases of six, all with the same label; in a six-bottle Solvent Pack (one each labeled acetone, deionized water, ethanol, isopropanol, methanol and universal); or six-bottle Cleaning Pack (one each labeled acetone, bleach, deionized water, saline, soap and universal bottles).

TRUSTED_SUSTAINABILITY

Specifications

Capacity 17 oz., 500mL
Capacity (Metric) 500 mL
Capacity (English) 17 oz.
Color Green
Diameter (English) 2.99 in.
For Use With (Application) Methanol
Includes Color-coded Silk screened labels, PTFE membrane
Material LDPE
Closure Color Green
Closure Included Yes
Autoclavable Not Autoclavable
Graduated No
Mouth Wide
Shape Cylindrical
Height (Metric) 173 mm
Product Type Wash Bottle
Height (English) 6.81 in.
Closure Material Polypropylene
Closure Size 38-415 mm
Diameter (Metric) 76 mm
Closure Type Polypropylene Closure
Through the Closure Dispensing No
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'