Learn More
Fisherbrand™ Easy-Squeeze Wash Bottles, 6/Cs.
Shop All Fisherbrand Products

Description
Includes
Come with colored polypropylene closures with PTFE membrane (except 02-897-2, which has an HDPE closure).
Easy to fill and ready to use
- Wide mouth
- Translucent and sputter-free
- With easy-to-read color-coded silk-screened labels
- Tips can be cut back to increase flow
- When not in use, bottles relieve internal pressure without dripping
Label information includes:
- Chemical name
- National Fire Protection Association (NFPA) codes and symbols
- Chemical Abstracts Service (CAS) number
- Hazard codes
- Laboratory protective equipment information
- Target organ information
- Recommended fill level
- LDPE recycling code
- Bottle volume capacity “For Dispensing Only, Not Recommended For Storage” statement
Not recommended for use above 60°C (140°F).
Order Info
Available in cases of six, all with the same label; in a six-bottle Solvent Pack (one each labeled acetone, deionized water, ethanol, isopropanol, methanol and universal); or six-bottle Cleaning Pack (one each labeled acetone, bleach, deionized water, saline, soap and universal bottles).
- Environmental benefits include:
- Renewable Energy

Specifications
Specifications
Capacity | 17 oz., 500mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 17 oz. |
Color | Blue |
Diameter (English) | 2.99 in. |
Includes | PTFE membrane |
Material | LDPE |
Closure Color | Blue |
Certifications/Compliance | ISO 13485:2003 |
Closure Included | Yes |
Show More |
Documents
Scientific Resources
Brochures
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% -
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.