missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Easy-Squeeze Wash Bottles, 6/Cs.
27.92 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Use promo code "23291" to get your promotional price.
Click to view available options
Color:
Assorted
Blue
Green
Natural
Red
Yellow
For Use With (Application):
Acetone
Bleach
Cleaning assortment pack
Deionized Water
Distilled Water
Ethanol
Isopropanol
Methanol
Saline
Soap
Solvent assortment pack
Universal
Water
Description
Includes
Come with colored polypropylene closures with PTFE membrane (except 02-897-2, which has an HDPE closure).
Easy to fill and ready to use
- Wide mouth
- Translucent and sputter-free
- With easy-to-read color-coded silk-screened labels
- Tips can be cut back to increase flow
- When not in use, bottles relieve internal pressure without dripping
Label information includes:
- Chemical name
- National Fire Protection Association (NFPA) codes and symbols
- Chemical Abstracts Service (CAS) number
- Hazard codes
- Laboratory protective equipment information
- Target organ information
- Recommended fill level
- LDPE recycling code
- Bottle volume capacity “For Dispensing Only, Not Recommended For Storage” statement
Not recommended for use above 60°C (140°F).
Order Info
Available in cases of six, all with the same label; in a six-bottle Solvent Pack (one each labeled acetone, deionized water, ethanol, isopropanol, methanol and universal); or six-bottle Cleaning Pack (one each labeled acetone, bleach, deionized water, saline, soap and universal bottles).
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Capacity | 17 oz., 500mL |
Capacity (Metric) | 500 mL |
Capacity (English) | 17 oz. |
Color | Assorted |
Diameter (English) | 2.99 in. |
For Use With (Application) | Solvent assortment pack |
Material | LDPE |
Certifications/Compliance | ISO 13485:2003 |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'