missing translation for 'onlineSavingsMsg'
Learn More
Fisherbrand™ Easy-Squeeze Wash Bottles, 6/Cs. Product Code.: 11785233
In Stock
to see stock.
Add to basket

Fisherbrand™ Easy-Squeeze Wash Bottles, 6/Cs.
GREENER_CHOICE

Product Code. 11785233
Natural, Ethanol
Click to view available options
Color:
Assorted
Blue
Green
Natural
Red
Yellow
Chemical Label:
Acetone
Assorted Cleaning
Assorted Solvents
Deionized Water
Distilled Water
Ethanol
Isopropanol
Methanol
Saline
Soap
Sodium Hypochlorite (Bleach)
Universal
Water
Wash bottles in this range that are prelabelled for intended usage with specific hazardous substances are also prelabelled with the appropriate GHS hazard warning symbols.
 

Product Code. 11785233

In Stock
to see stock.
Edge
Add to basket
Add to basket
Wash bottles in this range that are prelabelled for intended usage with specific hazardous substances are also prelabelled with the appropriate GHS hazard warning symbols.
 

Includes

Come with colored polypropylene closures with PTFE membrane (except 02-897-2, which has an HDPE closure).

Easy to fill and ready to use

  • Wide mouth
  • Translucent and sputter-free
  • With easy-to-read color-coded silk-screened labels
  • Tips can be cut back to increase flow
  • When not in use, bottles relieve internal pressure without dripping

Label information includes:

  • Chemical name
  • National Fire Protection Association (NFPA) codes and symbols
  • Chemical Abstracts Service (CAS) number
  • Hazard codes
  • Laboratory protective equipment information
  • Target organ information
  • Recommended fill level
  • LDPE recycling code
  • Bottle volume capacity “For Dispensing Only, Not Recommended For Storage” statement

Not recommended for use above 60°C (140°F).

Order Info

Available in cases of six, all with the same label; in a six-bottle Solvent Pack (one each labeled acetone, deionized water, ethanol, isopropanol, methanol and universal); or six-bottle Cleaning Pack (one each labeled acetone, bleach, deionized water, saline, soap and universal bottles).

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Capacity 17 oz., 500mL
Capacity (Metric) 500 mL
Capacity (English) 17 oz.
Color Natural
Diameter (English) 2.99 in.
Includes Color-coded Silk screened labels, PTFE membrane
Material LDPE
Closure Color Natural
Certifications/Compliance ISO 13485:2003
Closure Included Yes
Autoclavable Not Autoclavable
Graduated No
Mouth Wide
Shape Cylindrical
Height (Metric) 173 mm
Product Type Wash Bottle
Height (English) 6.81 in.
Chemical Label Ethanol
Closure Material Polypropylene
Closure Size 38-415 mm
Diameter (Metric) 76 mm
Closure Type Polypropylene Closure
Through the Closure Dispensing No
Show More Show Less
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Days

    26

    Hours

    13

    Minutes

    39

    Seconds

    59
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Save up to 94% on Teaching Lab Essentials
    Promo Code25721

    Expires2025-05-01

    New Offer
    Save Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Easy-Squeeze Wash Bottles, 6/Cs. >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.