missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 96-Well Plates

209.08 GBP valid until 2025-03-14
Use promo code "24150" to get your promotional price.
Description
For EIA and other applications
- Clear polystyrene
- Results with CV of <5% between wells and replicate values within ±10% of the mean Background of wells is within 0.005 absorbance units from mean Small lip at each well top and ridges between wells prevent well-to-well contamination Flat well bottom surface provides greater area for binding
- Rows and columns are labeled alphanumerically; plates stack for easy storage
- Specially treated to ensure high binding of hydrophobic or partially hydrophobic molecules like viral antigens and proteins
Order Info
Nonsterile microplates are supplied in bulk packs of 50 plates. Sterile microplates come individually wrapped with lids in packs of 40 plates.
Certifications
Cat. No. 21-377-203 is supplied with a certificate attesting to its high-binding capabilities.

Specifications
Certified | |
Without Lid | |
50 Plates, 40 Plates | |
Clear | |
Polystyrene | |
96 | |
High Binding | |
EIA and other applications |
Standard | |
8 x 12 | |
High Binding | |
96-well Assay Microplate | |
450 μL | |
Non-sterile | |
Rounded Square |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction