missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 96-Well Polypropylene Microplates

Product Code. p-156247 Shop All Fisherbrand Products
Click to view available options
Color:
Blue
Natural
Red
Yellow
Unit Size:
Pack of 80

Product Code. 11907954

Only null left
Add to basket
Add to basket

Ideal for storage—feature generous 500μL well volumes

  • Polypropylene
  • Minimum binding
  • Resistant to many reagents
  • Withstand temperatures from -80° to +121°C (-112° to +250°F)
  • Automation-friendly design optimized for HTS

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Array 8 x 12
Color Natural
Material Polypropylene
No. of Wells 96
Sterility Non-sterile
Well Shape Round
For Use With (Application) For storage feature generous 500μL well volumes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.