missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Absorption Macro Rectangular Cells with PTFE Lid

Fit standard spectrophotometer cell holders

£77.70 - £1035.00

Specifications

Type Macro Absorption Cell
Closure Material PTFE
Width (Metric) Interior 10mm
For Use With (Equipment) Standard Spectrophotometer Cell Holder
Width (Metric) 12.5 mm
View More Specs

Includes: PTFE lid

Products 15
Product Code Brand Capacity (Metric) Material Path Length Wavelength Range Price Quantity & Availability  
Product Code Brand Capacity (Metric) Material Path Length Wavelength Range Price Quantity & Availability  
15265953
View Documents
Fisherbrand™
14958101
1.75 mL Quartz 5 mm 200 to 2500 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15255953
View Documents
Fisherbrand™
14958100
3.5 mL Quartz 1 mm 200 to 2500 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15266013
View Documents
Fisherbrand™
14958110
3.5 mL Quartz 1 mm 200 to 2500 nm
£442.00
Pack of 2
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
 
15276013
View Documents
Fisherbrand™
14958111
1.5 mL Quartz 5 mm 200 to 2500 nm
£367.00
Pack of 2
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
 
15296013
View Documents
Fisherbrand™
14958113
3.5 mL Quartz 10 mm 200 to 2500 nm
£611.00
Pack of 4
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
 
15226023
View Documents
Fisherbrand™
14958115
0.35 mL Quartz 1 mm 200 to 3500 nm
£568.00
Pack of 2
Estimated Shipment: 03-04-2025
to see stock.
Add to basket
 
15286013
View Documents
Fisherbrand™
14958112
3.5 mL Quartz 10 mm 200 to 2500 nm
£1035.00
Pack of 2
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
 
15256023
View Documents
Fisherbrand™
14958118
3.5 mL Quartz 10 mm 200 to 3500 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15246023
View Documents
Fisherbrand™
14958116
3.5 mL Quartz 10 mm 200 to 3500 nm
£392.00
Pack of 2
Estimated Shipment: 03-04-2025
to see stock.
Add to basket
 
15266023
View Documents
Fisherbrand™
14958119
3.5 mL Special Optical Glass 10 mm 320 to 2500 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15276023
View Documents
Fisherbrand™
14958120
3.5 mL Optical Glass 10 mm 360 to 2500 nm
£126.00
Pack of 2
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
 
15286023
View Documents
Fisherbrand™
14958121
3.5 mL Optical Quartz 10 mm 260 to 2500 nm
£320.00
Pack of 2
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
 
15206033
View Documents
Fisherbrand™
14958123
3.5 mL Optical Glass 10 mm 360 to 2500 nm
£392.00
Pack of 4
Estimated Shipment: 12-06-2025
to see stock.
Add to basket
 
15216033
View Documents
Fisherbrand™
14958124
3.5 mL Optical Quartz 10 mm 260 to 2500 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15296023
View Documents
Fisherbrand™
14958122
3.5 mL Special Optical Glass 10 mm 320 to 2500 nm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Fit standard spectrophotometer cell holders

  • All cells are 12.5mm wide
  • 1, 5 or 10mm path length rectangular cells
  • Available with a variety of window materials for wide range of assays
  • Available in four materials e.g. Quartz, Optical Quartz, Special Optical Glass and Optical Glass
  • Quartz windows for transmission from far ultraviolet through the infrared
  • Glass windows for routine work in ultraviolet region and visible region

Specifications

Specifications

Macro Absorption Cell
10mm
12.5 mm
PTFE
Standard Spectrophotometer Cell Holder
Rectangle
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.