missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Absorption Quartz Micro Rectangular Cells with PTFE Lid

Fit standard spectrophotometer cell holders

£179.99 - £343.00

Specifications

Type Micro Absorption Cell
Material Quartz
Closure Material PTFE
Width (Metric) Interior 2mm
Includes 2 clear windows
View More Specs

Includes: PTFE lid

Products 2
Product Code Brand Capacity (Metric) Type Height (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Type Height (Metric) Price Quantity & Availability  
15256073
View Documents
Fisherbrand™
14958133
300 μL Micro Absorption Cell 25 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15266083
View Documents
Fisherbrand™
14958134
500 μL Micro Absorption Cell 45 mm
£343.00
Pack of 2
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
Description

Description

Fit standard spectrophotometer cell holders

  • All cells are 12.5mm wide
  • 10mm path length rectangular cells
  • Reduce reagent use with Micro cells
  • Quartz windows for transmission from far ultraviolet through the infrared

Specifications

Specifications

Micro Absorption Cell
PTFE
2 clear windows
10 mm
Standard Spectrophotometer Cell Holder
Transmission from far ultraviolet through the infrared
Quartz
2mm
2/Pk., 8 Pk./Cs.
200 to 2500 nm
12.5 mm
Rectangle
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.