missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Semi-Micro Cuvette with PTFE lid
Contains quartz windows for transmission from far ultraviolet through the infrared.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 0.01000kg
Includes: PTFE lid
Description
- Reduces the use of reagent with micro cells
- Contains two clear windows made of quartz
- With PTFE lid
Specifications
Semi-micro Cuvette | |
Quartz | |
4mm | |
10 mm | |
49.5 mm | |
12.5 mm | |
Rectangle |
1500 μL | |
PTFE | |
Ea. | |
200 to 2500 nm | |
Standard spectrophotometer | |
Transmission from far ultraviolet through the infrared |