missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Absorption Ultra Micro Rectangular Cells with PTFE Lid
Fit standard spectrophotometer cell holders
£397.90 - £481.00
Specifications
Type | Ultra-micro Absorption Cell |
---|---|
Material | Quartz |
Closure Material | PTFE |
Width (Metric) Interior | 2mm |
Quantity | Ea., 8 Ea./Cs. |
Includes: PTFE lid or stopper
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Description | Price | Quantity & Availability | ||||
15296103
|
Fisherbrand™
|
100 μL | Center:15mm |
£481.00
Each |
Estimated Shipment: 18-10-2024
Log in to see stock. |
||||
15246103
|
Fisherbrand™
|
100 μL | Center:8.5mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15216123
|
Fisherbrand™
|
50 μL | Center:8.5mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15286123
|
Fisherbrand™
|
50 μL | Center:15mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- All cells are 12.5mm wide
- 1, 5 or 10mm path length rectangular cells
- Reduce reagent use with Ultramicro cells
- Quartz windows for transmission from far ultraviolet through the infrared
- Glass windows for routine work in ultraviolet region and visible region
Specifications
Ultra-micro Absorption Cell | |
PTFE | |
Ea., 8 Ea./Cs. | |
200 to 2500 nm | |
Standard spectrophotometer cell holder | |
Transmission from far ultraviolet through the infrared |
Quartz | |
2mm | |
10 mm | |
45 mm | |
12.5 mm | |
Rectangle |