Learn More
Fisherbrand™ Aluminum Foil, Standard-Gauge Roll
For sealing, weighing, and transferring samples
Brand:  Fisherbrand™ 01213103
 View more versions of this product
Product Code. 11779408
  • £9.75 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Ideal for weighing, sealing, and tranferring your samples.
  • Packed in individual dispenser box with cutter bar
  • Heavy-gauge rolls offer greater folding strength, good for covering sharp edges
Specifications
Specifications
Sealing, Weighing, Transferring Samples
Foil
46cm
Aluminum
25 ft.
18 in.
7.6 m
24 μm