missing translation for 'onlineSavingsMsg'
Learn More

Bio Plas™ Astral™ Inoculation Device System

Product Code. 11534000 Shop All Thermo Scientific Products
Click to view available options
Product Type:
Inoculating Loop
Color:
Yellow
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11534000

Brand: Bio Plas™ 7000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Autoclavable handle with every case

Economical, disposable inoculation system designed for safety and dependable sterility

Economical, disposable inoculation system designed for safety and dependable sterility

Calibrated using Evans Blue Dye spectrophotometric procedure and gravimetric method

  • Free of DNA, RNase, DNase, pyrogens and PCR inhibitors
  • Astral loops, needles, and Clone-Piks are packaged static free into individual tubular chambers, which are then hermetically sealed with aluminum film
  • A dust cover is heat-shrunk onto the unit before it is irradiation sterilized
  • Use on an individual basis, or for quantitative procedures such as sampling, urine counts, or serial dilutions

To use:

  • Puncture aluminum film with the handle, pushing down until it attaches to the loop, needle, or Clone-Pik
  • Withdraw handle; inoculation device is ready for many applications

Order Info

Supplied 10 racks of 96 to a case

TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Volume 1 μL
Sterility Sterile
Color Yellow
Product Type Inoculating Loop
For Use With (Equipment) Astral™ inoculation device system
Includes Autoclavable handle
Packaging Type Rack
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt