missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Autoclavable Biohazard Waste Bags
Dual tested compliant.
Brand: Fisherbrand™
Additional Details : Weight : 1.00000kg
Description
Dual tested compliant
Autoclaving is a very common way to sterilize infectious waste through the use of high temperature, high pressure steam over a specific time period. Sterilized waste is less expensive to dispose. Dual-tested autoclave bags are ASTM D1922 Elmendorf Tear tested to exceed 480g of tear strength (MD and TD) and ASTM D1709 tested to exceed 165g dart drop strength.- Integral steam processing indicator that turns brown when autoclaved
- Bags comply with US DOT 49CFR Sections 173-197 and specific state regulations
- Operating temperature tolerance up to 285degreeFdegreeAvailable in various sizes
- Bag with black print
- Gauge: 2mil
Specifications
Autoclavable | |
Red with Black Print | |
58.4 cm |
7 to 10 gal. | |
23 x 19 in. (58.4 x 48.3 cm) | |
48.03 cm |