Learn More
Fisherbrand™ Disposable Shoe Cover
Spunbond polypropolyene shoe covers protect against dirt, grime and certain dry particulates
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 1.80000kg
Descripción
Spunbond polypropolyene shoe covers protect against dirt, grime and certain dry particulates
- Single-use
- Blue
- Polypropylene
- Latex-free
- Without anti-skid sole
- Waterproof
Especificaciones
Universal | |
Yes | |
Single-use | |
Without anti-skid tread |
Polypropylene | |
Blue | |
Standard |