missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Bitran™ Specimen Storage Bags

Choice of Saranex™ odor-control film for long-term storage or polyethylene for short-term storage

Brand:  Fisherbrand™

Additional Details : Weight : 0.45000kg

 View more versions of this product

Product Code. 16488115

  • £182.00 / Pack of 250
Estimated Shipment: 20-08-2024
to see stock.
Add to basket

Description

Description

Choice of Saranex™ odor-control film for long-term storage or polyethylene for short-term storage

  • Use for collecting, transporting and storing tissue specimens
  • Minimize storage space and help to eliminate leakage, breakage and contamination
  • Resealable double-lock zipper closure
  • Designed for storage of tissues and fixatives and for use as mini-anaerobic environments

Specifications

Specifications

Zip Seal
250
2 in.
4 in.
Saranex Film
2 x 4 in. (5.1 x 10.2cm)
5.1 cm
10.2 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions