Learn More
Fisherbrand™ BloodBloc™ Biohazard Wipes, Liners and Rolls
Prevent leakage of fluids and chemicals to hands, gloves and work surfaces
Brand:  Fisherbrand™ 0667054
 View more versions of this product
Product Code. 16207050
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Wipes and Liners are non-sterile and not intended for use on patients.
  • Soft, white absorbent wipe with bright-orange polyethylene backing is impermeable to blood, body fluids and strong solvents including xylene, xylene substitutes, acetones, ketones, alcohols and formalin
  • Nonwoven cellulose captures and retains fluid; nonlinting for use with delicate instrumentation such as blood counters
  • Orange flexible impermeable plastic backing prevents soak-through of pathogens and chemicals to gloves which help minimize cross-contamination when lab personal open doors or touch surfaces
  • Can also be used as mats and liners; rolls can be cut to desired length
  • One time use only
Specifications
Specifications
Biohazard Roll
30.48 m
40.64 cm
Orange backing
100 ft. x 16 in. (30.48 m x 40.64 cm)
Bench tops, specimen collection tubs, trays, fume hoods, Bio-Safety and chemical cabinets
100 ft.
16 in.
Heavy
Single-use
Prevent soak through of blood, body fluids, tissue and chemicals to gloves and surfaces when handling tubes, specimens, plates.
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Product Certifications
Ofertas especiales
Ofertas especiales