missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ BloodBloc™ Biohazard Wipes, Liners and Rolls
Prevent leakage of fluids and chemicals to hands, gloves and work surfaces
Brand: Fisherbrand™ 0667055
61.83 GBP valid until 2024-12-31
Use promo code "24046" to get your promotional price.
Description
Wipes and Liners are non-sterile and not intended for use on patients.
- Soft, white absorbent wipe with bright-orange polyethylene backing is impermeable to blood, body fluids and strong solvents including xylene, xylene substitutes, acetones, ketones, alcohols and formalin
- Nonwoven cellulose captures and retains fluid; nonlinting for use with delicate instrumentation such as blood counters
- Orange flexible impermeable plastic backing prevents soak-through of pathogens and chemicals to gloves which help minimize cross-contamination when lab personal open doors or touch surfaces
- Can also be used as mats and liners; rolls can be cut to desired length
- One time use only
Specifications
Biohazard Roll | |
30.48 m | |
40.64 cm | |
Orange backing | |
100 ft. x 16 in. (3048 x 40.64 cm) | |
Bench tops, specimen collection tubs, trays, fume hoods, Bio-Safety and chemical cabinets |
100 ft. | |
16 in. | |
Standard | |
Single-use | |
Prevent soak through of blood, body fluids, tissue and chemicals to gloves and surfaces when handling tubes, specimens, plates. |