missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ BloodBloc™ Biohazard Wipes, Liners and Rolls
Prevent leakage of fluids and chemicals to hands, gloves and work surfaces
£10.10 - £174.00
Specifications
Disposable | Single-use |
---|---|
For Use With (Application) | Prevent soak through of blood, body fluids, tissue and chemicals to gloves and surfaces when handling tubes, specimens, plates. |
Product Code | Brand | Absorbancy | Length (Metric) | Width (Metric) | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Absorbancy | Length (Metric) | Width (Metric) | Product Type | Price | Quantity & Availability | |||||
16207040
|
Fisherbrand™
06-670-36 |
Super | 7.62 cm | 7.62 cm | Biohazard Wipe | N/A | ||||||
15505895
|
Fisherbrand™
066701 |
Standard | 7.62 cm | 7.62 cm | Biohazard Wipe |
£18.93
Pack of 250 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||||
16217040
|
Fisherbrand™
0667038 |
Super | 55.88 cm | 40.64 cm | Biohazard Liner |
£174.00
Pack of 100 |
Estimated Shipment: 22-05-2025
Log in to see stock. |
|||||
16237040
|
Fisherbrand™
06-670-48 |
Super | 30.48 m | 40.64 cm | Biohazard Roll | N/A | ||||||
16247040
|
Fisherbrand™
0667049 |
Heavy | 7.62 cm | 7.62 cm | Biohazard Wipe |
£13.15
Pack of 200 |
Estimated Shipment: 19-05-2025
Log in to see stock. |
|||||
16277040
|
Fisherbrand™
0667051 |
Heavy | 22.86 cm | 20.32 cm | Biohazard Wipes | N/A | ||||||
16267040
|
Fisherbrand™
0667050 |
Heavy | 10.16 cm | 10.16 cm | Biohazard Wipe | N/A | ||||||
16207050
|
Fisherbrand™
0667054 |
Heavy | 30.48 m | 40.64 cm | Biohazard Roll | N/A | ||||||
16217050
|
Fisherbrand™
0667055 |
Standard | 30.48 m | 40.64 cm | Biohazard Roll |
£87.85
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
16297040
|
Fisherbrand™
0667053 |
Heavy | 40.64 cm | 40.64 cm | Biohazard Liner | N/A | ||||||
Description
Wipes and Liners are non-sterile and not intended for use on patients.
- Soft, white absorbent wipe with bright-orange polyethylene backing is impermeable to blood, body fluids and strong solvents including xylene, xylene substitutes, acetones, ketones, alcohols and formalin
- Nonwoven cellulose captures and retains fluid; nonlinting for use with delicate instrumentation such as blood counters
- Orange flexible impermeable plastic backing prevents soak-through of pathogens and chemicals to gloves which help minimize cross-contamination when lab personal open doors or touch surfaces
- Can also be used as mats and liners; rolls can be cut to desired length
- One time use only
Specifications
Single-use | |
Prevent soak through of blood, body fluids, tissue and chemicals to gloves and surfaces when handling tubes, specimens, plates. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ BloodBloc™ Biohazard Wipes, Liners and Rolls