missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sterile Cell Strainers

Product Code. 11517532 Shop All Fisherbrand Products
Click to view available options
Mesh Size:
40 μm
70 μm
100 μm
Color:
Blue
White
Yellow
Unit Size:
Pack of 50

Product Code. 11517532

Only null left
Add to basket
Edge
Add to basket

Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50mL conical tubes. Available in three color-coded mesh sizes.

Rapidly isolate primary cells for single-cell suspensions using Fisherbrand™ Cell Strainers. Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50mL conical tubes. Available in three color-coded mesh sizes.

Rapidly isolate primary cells
  • Fast, simple alternative to gauze filtration
  • Reliably dissociates cells from clumps or primary tissues
  • Consistently obtain a uniform single-cell suspension
  • Ideal for preparation of:
    • Single-cell suspensions of stem and primary (blood, tissue, cancer) cells
    • Specimens for primary cell cultures and immunogens
    • Freezing stocks
    • Filtering agglutinative proteins produced in inactivation serum
Reliable and easy to use
  • Strong nylon mesh with evenly spaced pores for consistent results
  • Sterilized by gamma irradiation and non-cytotoxic to maintain sample integrity
  • Includes an extended lip for easier handling with forceps
  • Color-coded polypropylene frame with tab enables easy handling and identification
  • Individually packed, ready to use and disposable

Specifications

Mesh Material Nylon
Disposable Single-use
Color Yellow
Material Polypropylene
Product Type Cell Strainer
Sterility Sterile
For Use With (Equipment) Conical Tubes, 50 mL
Mesh Size 100 μm
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis