missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Sterile Cell Strainers
Shop All Fisherbrand Products
Click to view available options
Mesh Size:
40 μm
70 μm
100 μm
Color:
Blue
White
Yellow
Förpackningsstorlek
Pack of 50
Beskrivelse
Rapidly isolate primary cells for single-cell suspensions using Fisherbrand™ Cell Strainers. Sterile and ready to use for straining stem and primary cell samples. Disposable strainers feature universal fit for compatibility with most 50mL conical tubes. Available in three color-coded mesh sizes.
Rapidly isolate primary cells
- Fast, simple alternative to gauze filtration
- Reliably dissociates cells from clumps or primary tissues
- Consistently obtain a uniform single-cell suspension
- Ideal for preparation of:
- Single-cell suspensions of stem and primary (blood, tissue, cancer) cells
- Specimens for primary cell cultures and immunogens
- Freezing stocks
- Filtering agglutinative proteins produced in inactivation serum
- Strong nylon mesh with evenly spaced pores for consistent results
- Sterilized by gamma irradiation and non-cytotoxic to maintain sample integrity
- Includes an extended lip for easier handling with forceps
- Color-coded polypropylene frame with tab enables easy handling and identification
- Individually packed, ready to use and disposable
Tekniske data
Tekniske data
| Mesh Material | Nylon |
| Disposable | Single-use |
| Color | White |
| Material | Polypropylene |
| Product Type | Cell Strainer |
| Sterility | Sterile |
| For Use With (Equipment) | Conical Tubes, 50 mL |
| Mesh Size | 70 μm |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion