Learn More
Fisherbrand™ Cheesecloth Miniwipes, 200 pk.
Each wipe is a folded 4 in. (10.2cm) square pad that unfolds to approximately 7 x 14 in.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 0.28440kg
Description
Fisherbrand™ Miniwipes can be used on microscopes, optical surfaces, glassware, pH electrodes. Each wipe is a folded 4 in. (10.2cm) square pad that unfolds to approximately 7 x 14 in.
- Certified 100% reagent-grade cotton
- Will not scratch sensitive surfaces
- Absorbs 6 times weight
- Size: 7 x 14 in. folded to 4 in. sq.
Order Info
Box dimensions: 4.5L x 4.5W x 8 in. H (11 x 11 x 20cm).
Specifications
Cheesecloth Miniwipe | |
White | |
For use to wipe cuvettes and microscopes, clean cameras, and dry delicate Glassware, syringes, pipettes, and pH electrode |
Cotton | |
7 x 14 in. | |
Microscopes, optical surfaces, Glassware, pH electrodes |