missing translation for 'onlineSavingsMsg'
Learn More
Fisherbrand™ Clear Straight Sided Round Bottles without Cap Product Code.: 11553792

Fisherbrand™ Clear Straight Sided Round Bottles without Cap

Product Code. 11553792
480 mL
Click to view available options
Capacity (Metric):
480 mL
This item is not returnable. View return policy

Product Code. 11553792

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed without shoulders for maximum storage capacity and content visibility. Fisherbrand™ Clear Straight-Sided Jars without Cap are for multipurpose use.

Clear (Flint) Straight Sided Round Glass Jars are designed without shoulders for maximum storage capacity and content visibility. These multipurpose jars are very versatile. Our straight sided jars are available in a wide variety of sizes, from 1oz to 32oz. Also available in tall/slender style in 8oz and 16oz sizes.

  • Designed without shoulders for maximum storage capacity
  • Straight sided walls allow for the complete removal of contents
  • Available in a wide variety of sizes, from 1 to 32 oz.

Specifications

Capacity (Metric) 480 mL
Capacity (English) 16 oz.
Color Clear
Material Glass
Quantity 12/Cs.
Neck Finish 89-400
Closure Included No
Mouth Wide
Shape Straight-sided
Product Type Straight Sided Jar
Closure Size 89-400 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Clear Straight Sided Round Bottles without Cap >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.