Learn More
Fisherbrand™ Color Coders for Externally and Internally Threaded Cryogenic Storage Vials
For sample identification
Brand:  Fisherbrand™
54.19 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 46
Additional Details : Weight : 0.08400kg
 View more versions of this product
Product Code. 11937974
  • £74.56 / Pack of 500
Estimated Shipment: 14-10-2024
to see stock.
Add to basket
Description
Description
Fit into the caps of Fisherbrand Externally Threaded Cryogenic Storage Vials (10-500-25, -26, and -27) or Internally Threaded Cryogenic Storage Vials (12-567-500, -501, -502).
  • For color-coded sample identification
  • Polystyrene
  • Nonsterile
Specifications
Specifications
Yellow
Polystyrene
Non-sterile
For externally threaded vials