Learn More
Fisherbrand™ Combs for Midi-Plus Wide Format Horizontal Gel Units, 2mm Thick
Description
- Recommended to scale-up nucleic acid volumes for preparatory techniques
- Maximises sample volume
- Height-adjustable without any requirement for specialist tools or comb holders, to give user full control over well depth and sample-loading volume
- Rigid comb back prevents heat-induced warping
- Reversible loading guides sit directly above each well to provide a convenient loading template for single- and multi-channel pipets
- 12-sample, 25-sample and 50-sample combs are Multichannel pipettor compatible
Specifications
Blue | |
Comb | |
1 Each | |
30 μL |
Midi-Plus Wide Format Horizontal Gel Units | |
50 | |
2 mm |