missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Power Adapters for Fisherbrand Digital Thermometers

Accessory for use with digital thermometers.

Brand:  Fisherbrand™

Code : NEW

Additional Details : Weight : 0.13500kg

Product Code. 15826741

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Accessory for use with digital thermometers.

Switch meter operation from battery to 115V 60Hz electrical power.
Specifications

Specifications

Switch meter operation from battery
Easy Use Adaptor
Digital Thermometer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers