missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Disposable Cuvettes
Choice of polystyrene or methacrylate
£79.00 - £332.00
Specifications
Disposable | Yes |
---|---|
Length (Metric) | 12mm |
Quantity | 500 Case |
Path Length | 10 mm |
Height (Metric) | 45 mm |
Product Code | Brand | Capacity (Metric) | Type | Material | Wavelength Range | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Type | Material | Wavelength Range | Price | Quantity & Availability | ||
11954395
|
Fisherbrand™
|
4.5 mL | Cuvette | Polystyrene | 340 to 750 nm |
£260.06
Pack of 500 |
In Stock
Log in to see stock. |
||
11944385
|
Fisherbrand™
|
4.5 mL | Standard Cuvette | PMMA | 285 to 750 nm |
£217.75
Pack of 500 |
In Stock
Log in to see stock. |
||
11924405
|
Fisherbrand™
|
4.5 mL | Cuvette | PMMA | 285 to 750 nm |
£332.00
Pack of 500 |
In Stock
Log in to see stock. |
||
11682599
|
Fisherbrand™
|
4.5 mL | Standard Cuvette | Polystyrene | 340 to 750 nm |
£110.00
Pack of 500 |
In Stock
Log in to see stock. |
||
11602609
|
Fisherbrand™
|
1.5 mL | Semi-micro Cell | Polystyrene | 340 to 750 nm |
£79.00
Pack of 500 |
In Stock
Log in to see stock. |
||
11904385
|
Fisherbrand™
|
1.5 mL | Semi-micro Cell | Methacrylate | 285 to 750 nm |
£120.74
Pack of 500 |
In Stock
Log in to see stock. |
||
Description
Fisherbrand™ Disposable Cuvettes are available in choice of polystyrene or methacrylate. Select polystyrene cuvets for assays in visible spectral range (340 to 750nm); for accuracy throughout UV/Vis range (285 to 750nm), choose cuvets made from a superior grade of methacrylate.
- Disposable 10mm pathlength cuvets designed for visible and UV/Vis assays
- Choice of 4.5 and 1.5mL sample capacity
- Clear four-sided cuvets are ideal for fluorimetry and nephelometry, as well as spectrophotometry
- Scale down assay volumes and save on reagents by using semimicro cuvets with capacity to 1.5mL
- Molded-in arrow indicates direction of transmission on standard and semimicro cuvets—helping assure uniformity
- Each case contains cuvets with same mold impression number—assuring precision between samples—standard deviation is <±0.005 extinction units between cuvets
Order Info
Packaged 100 cuvets per stackable tray; five trays to a case. Outside dimensions: 0.47L x 0.47W x 1.77 in. H (12 x 12 x 45mm).
Specifications
Yes | |
500 Case | |
45 mm | |
12 mm |
12mm | |
10 mm | |
Spectrophotometers, Fluorometers and Nephelometers | |
For assays in visible spectral range, accuracy throughout UV/Visible range |