missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Disposable Petri Dishes

Tight-fitting lid prevents dehydration

Brand:  Fisherbrand™

Additional Details : Weight : 1.50000kg

 View more versions of this product

Product Code. 16640272

  • £104.00 / Pack of 150
Estimated Shipment: 28-08-2024
to see stock.
Add to basket

Description

Description

Secure, tight-fitting lid helps to prevent dehydration.

Sterilized Petri Dishes

  • 47mm dishes can be used with solid agar medium or add absorbent pads for liquid medium
  • Molded polystyrene
Sterilized Pad Dishes
  • Preloaded 47mm dishes with sterilized absorbent pads
  • Minimize contamination
  • Pure cellulose pad
  • Extended 3-year shelf-life

 

Specifications

Specifications

Single-use
Round
Petri Dish without Pad
47 mm
Polystyrene
Sterile
150 Pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers