missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Elite™ Variable Volume, Multichannel Pipettes
GREENER_CHOICE

Product Code. 11855772 Shop All Fisherbrand Products
Click to view available options
Volume (Metric):
1 to 10 μL
10 to 100 μL
30 to 300 μL
5 to 50 μL
Accuracy:
1 to 4.6%
1.3 to 5%
1.5 to 5%
2.4 to 12%
Number of Channels:
8
12
Unit Size:
Each

Product Code. 11855772

Only null left
Add to basket
Add to basket

New, improved ergonomic design provides newly designed finger rest and index finger and hand support for maximum comfort

New, improved ergonomic design provides newly designed finger rest and index finger and hand support for maximum comfort

  • Fully autoclavable - entire pipetter can be steam autoclaved
  • Extremely low plunger forces - work longer without fatigue and reduce the risk of RSI
  • Soft-touch tip ejection—a gearing mechanism reducing tip ejection force up to 50%
  • Ergonomic comfortable lightweight handle - comfortable grip and effortless tip attachment
  • Optimal fit with SureOne pipetter tips - best pipetting performance
  • Super blow-out piston - creates 150% bigger air push than conventional piston for delivery of the micro size drops

TRUSTED_SUSTAINABILITY

Specifications

Accuracy 2.4 to 12%
Compatible Tips SureOne™ pipetter tips
Autoclavable Autoclavable
For Use With SureOne Pipette Tip
Number of Channels 12
Product Line Elite
Type Variable Volume Pipette
Volume (Metric) 1 to 10 μL
Finger Style Ergonomic
Increments 0.02μL
Warranty 3 Years
Show More Show Less

As part of our commitment to continuous product improvement, your delivery may contain products with some minor color variation, but their form, fit and function will not be affected. Any questions contact your sales representative.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.