missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Elite™ Variable Volume, Multichannel Pipettes
GREENER_CHOICE

New, improved ergonomic design provides newly designed finger rest and index finger and hand support for maximum comfort

Brand:  Fisherbrand™ 4682050

321.75 GBP valid until 2024-12-31
Use promo code "21925" to get your promotional price.


 View more versions of this product

Product Code. 11875772

  • £462.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

New, improved ergonomic design provides newly designed finger rest and index finger and hand support for maximum comfort

  • Fully autoclavable - entire pipetter can be steam autoclaved
  • Extremely low plunger forces - work longer without fatigue and reduce the risk of RSI
  • Soft-touch tip ejection—a gearing mechanism reducing tip ejection force up to 50%
  • Ergonomic comfortable lightweight handle - comfortable grip and effortless tip attachment
  • Optimal fit with SureOne pipetter tips - best pipetting performance
  • Super blow-out piston - creates 150% bigger air push than conventional piston for delivery of the micro size drops

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1.5 to 5%
Autoclavable
12
Variable Volume Pipette
Ergonomic
SureOne™ Pipetter tips
SureOne™ Pipette Tip
Elite™
5 to 50 μL
0.1μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.