Learn More
Fisherbrand™ Erlenmeyer Flask Clamps for Ultrasonic Units
Safely and securely attach Erlenmeyer flasks to Fisherbrand™ ultrasonic cleaning units. Fisherbrand™ Erlenmeyer Flask Clamps are manufactured using strong and durable stainless steel for superior stability.
Brand:  Fisherbrand™ 1007557
 View more versions of this product
Product Code. 10084622
  • £31.30 / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Safe and secure attachment
  • Manufactured from strong and durable stainless steel
  • Provides superior stability
Specifications
Specifications
Flask Clamp
Ultrasonic Cleaner
500 mL Erlenmeyer Flask