Learn More
Fisherbrand™ Finnpipette™ II Digital Multichannel Pipetters
Advanced pipetting performance in a lightweight, ergonomic design
Brand:  Fisherbrand™
Code : 46
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11827511
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • 8- and 12-channel pipetters, ideal for multiplate applications
  • Minimum force allows tips to be ejected with just a gentle push
  • Reverse pipetting mode allows pipetting of viscous and surface-active solutions
  • Made from inert, biologically inactive polymers
  • Easy to disassemble for routine maintenance and cleaning
  • Serial number and calibration certificate included
  • Easy, in-lab calibration
Superior Design
  • Rounded hand grip and modified grippy finger rest ensure secure grip and correct pipetting angle; help minimize hand fatigue
  • Large digital display is easy to read, even when pipetting
  • Large color-coded knob makes it easy to set and identify desired volume; fast-dial system allows 0.1μL fine adjustment
  • Separate soft-touch tip ejector relieves hand stress, prevents accidental detipping
  • Super blowout feature on micro models ensures highest level of accuracy and precision at low-volume settings
Order Info
For optimum performance, use Fisherbrand SureOne™ Pipet Tips or other universal fit pipet tips.
Services
Three years
Specifications
Specifications
Autoclavable
8
Digital Pipetter
5μL
Yellow
Finnpipette™ II
50 to 300 μL
3 Years