missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Flat and Domed 8-Cap Strips, Flat Cap Strips
Suitable for use with 96-well PCR plates and PCR tube strips
£45.90 - £47.85
Specifications
Material | Polypropylene |
---|---|
Color | Natural |
For Use With (Equipment) | 96-well PCR Plates or PCR Tube Strips |
Product Code | Brand | Format | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Price | Quantity & Availability | |||||
12184102
|
Fisherbrand™
14230230 |
Flat Cap Strips |
£45.90
Pack of 250 |
In Stock
Log in to see stock. |
|||||
11859251
|
Fisherbrand™
14230231 |
Domed Cap Strips |
£47.85
Pack of 250 |
In Stock
Log in to see stock. |
|||||
Description
Suitable for use with 96-well PCR plates and PCR tube strips
- Eight caps per strip
- Cleanroom produced
- Virgin polypropylene
- Flat caps offer large, labeled end tabs for easy removal and strip orientation
Specifications
Polypropylene | |
Natural | |
96-well PCR Plates or PCR Tube Strips |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title