missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Fluorescence Rectangular Cells: Micro Cells With PTFE Lids
Fit standard spectrophotometer cell holders
£473.70 - £766.00
Specifications
Type | Micro Fluorescence Cell |
---|---|
Material | Quartz |
Closure Material | PTFE |
Wavelength Range | 200 to 2500 nm |
Height (Metric) | 45 mm |
Includes: PTFE lid or stopper
Product Code | Brand | Type | Path Length | Capacity (Metric) | Description | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Path Length | Capacity (Metric) | Description | Price | Quantity & Availability | ||
15205993
|
Fisherbrand™
|
Micro Fluorescence Cell | 3 mm | 45 μL | Center H: 8.5mm |
£766.00
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
15266003
|
Fisherbrand™
|
Micro Fluorescence Cell | 1.5 mm | 12 μL | Center H: 8.5mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15296003
|
Fisherbrand™
|
Micro Fluorescence Cell | 1.5 mm | 12 μL | Center H: 15mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15235973
|
Fisherbrand™
|
Micro Fluorescence Cell | 10 mm | 100 μL | Center H: 8.5mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15235993
|
Fisherbrand™
|
Micro Fluorescence Cell | 3 mm | 45 μL | Center H: 15mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
15255973
|
Fisherbrand™
|
Micro Fluorescence Cell | 10 mm | 100 μL | Center H: 15mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- All cells are 12.5mm wide
- 1, 5 or 10mm path length rectangular cells
- Reduce reagent use with micro cells
- Wavelength range for all cells is 200 to 2500nm
- Quartz windows for transmission from far ultraviolet through the infrared
Specifications
Micro Fluorescence Cell | |
PTFE | |
45 mm | |
12.5 mm | |
Rectangle |
Quartz | |
200 to 2500 nm | |
Standard spectrophotometer cell holder | |
Transmission from far ultraviolet through the infrared |