missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PurSwab Foam Swabs

Flexible foam applicators. Tip is comprised of an open-cell polyurethane foam. Ideal for a variety of cleaning applications.

Brand:  Fisherbrand™ 149603L

51.61 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 16438115

  • £63.65 / Pack of 50
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

Flexible mini-tip foam applicators. Tip is comprised of an open-cell polyurethane foam. Ideal for a variety of cleaning applications.

Used in controlled environments, cleanrooms and industrial settings.  Coating rechargeable copier cartridges.  Applying solutions or solvents.  General cleaning swab.  Tip is thermally bonded to handle so there is no contaminating adhesives used.  Tip is lint and residue free.  Can be used with IPA and DI water or dry. Made in USA
Specifications

Specifications

50
0.245 in.
Coating rechargeable copier cartridges, applying solutions or solvents, general cleaning
0.622 cm
5.00 in.
0.375 in.
2.2cm
0.687 in.
Non-sterile
Polypropylene
Polyurethane Foam
ISO 9001:2008, ISO 13485:2003, FDA GMP's
5.17 in.
Swab
13.1cm
12.7 cm
0.89 in.
1.745 cm
Medical
Thermal
Rectangle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.